3-Chlor-4-(trifluormethoxy)benzonitril

Lieferant: Apollo Scientific
Achtung

PC0178-1G PC0178-5G PC0178-10G
APOSPC0178-1GEA 21.9 EUR
APOSPC0178-1G APOSPC0178-5G APOSPC0178-10G
3-Chlor-4-(trifluormethoxy)benzonitril
3-Chlor-4-(trifluormethoxy)benzonitril
Formel: C₈H₃ClF₃NO
Molecular Weight: 221.57 g/mol
Schmelzpunkt: 39…41 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD01631557
CAS-Nummer: 129604-26-8
UN: 3439
ADR: 6.1 III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR