3-Chlor-4-(trifluormethoxy)benzylbromid 98%

Lieferant: Apollo Scientific
Gefahr

PC0181-1G PC0181-5G PC0181-25G PC0181-100G
APOSPC0181-1GEA 0 EUR
APOSPC0181-1G APOSPC0181-5G APOSPC0181-25G APOSPC0181-100G
3-Chlor-4-(trifluormethoxy)benzylbromid 98%
3-Chlor-4-(trifluormethoxy)benzylbromid
Formel: C₈H₅BrClF₃O
Lagertemperatur: Raumtemperatur
MDL: MFCD01631560
CAS-Nummer: 261763-18-2
UN: 1760
ADR: 8,III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR