5-Chloro-2-(trifluoromethyl)benzoyl chloride
Lieferant: Apollo Scientific
|
Gefahr
|
PC0189-250MG
APOSPC0189-250MGEA
21.9
EUR
APOSPC0189-250MG
5-Chloro-2-(trifluoromethyl)benzoyl chloride
5-Chloro-2-(trifluoromethyl)benzoyl chloride
Formel:
C₈H₃Cl₂F₃O Siedepunkt: 214…217 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD01631354 CAS-Nummer: 320-84-3 UN: 3265 ADR: 8,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...