2-Chlor-3-(trifluormethyl)benzylalkohol 98%

Lieferant: Apollo Scientific

PC0190-5G PC0190-1G
APOSPC0190-5GEA 85.7 EUR
APOSPC0190-5G APOSPC0190-1G
2-Chlor-3-(trifluormethyl)benzylalkohol 98%
2-Chlor-3-(trifluormethyl)benzylalkohol
Formel: C₈H₆ClF₃O
Lagertemperatur: Raumtemperatur
MDL: MFCD01631537
CAS-Nummer: 261763-20-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR