5-Chlor-2-(trifluormethyl)benzylalkohol

Lieferant: Apollo Scientific
Achtung

PC0193-10G PC0193-1G PC0193-5G PC0193-25G
APOSPC0193-10GEA 406 EUR
APOSPC0193-10G APOSPC0193-1G APOSPC0193-5G APOSPC0193-25G
5-Chlor-2-(trifluormethyl)benzylalkohol
5-Chlor-2-(trifluormethyl)benzylalkohol
Formel: C₈H₆ClF₃O
Lagertemperatur: Raumtemperatur
MDL: MFCD01631355
CAS-Nummer: 261763-21-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR