4-Chlor-3-(trifluormethyl)benzylbromid 98%
Lieferant: Apollo Scientific
|
Gefahr
|
PC0194-100G
PC0194-25G
PC0194-5G
APOSPC0194-100GEA
213
EUR
APOSPC0194-100G
APOSPC0194-25G
APOSPC0194-5G
4-Chlor-3-(trifluormethyl)benzylbromid 98%
4-Chlor-3-(trifluormethyl)benzylbromid
Formel:
C₈H₅BrClF₃ Schmelzpunkt: 36…40 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD01631540 CAS-Nummer: 261763-23-9 UN: 3261 ADR: 8,II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...