5-Chlor-2-(trifluormethyl)benzylbromid 98%

Lieferant: Apollo Scientific
Gefahr

PC0195-10G PC0195-5G PC0195-1G
APOSPC0195-10GEA 0 EUR
APOSPC0195-10G APOSPC0195-5G APOSPC0195-1G
5-Chlor-2-(trifluormethyl)benzylbromid 98%
5-Chlor-2-(trifluormethyl)benzylbromid
Formel: C₈H₅BrClF₃
Lagertemperatur: Raumtemperatur
MDL: MFCD01631356
CAS-Nummer: 261763-24-0
UN: 3265
ADR: 8,II

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR