4-Chloro-3-(trifluoromethyl)cinnamic acid 97%

Lieferant: Apollo Scientific
Gefahr

PC0196-1G PC0196-5G PC0196-10G
APOSPC0196-1GEA 0 EUR
APOSPC0196-1G APOSPC0196-5G APOSPC0196-10G
4-Chloro-3-(trifluoromethyl)cinnamic acid 97%
4-Chloro-3-(trifluoromethyl)cinnamic acid
Formel: C₁₀H₆ClF₃O₂
Lagertemperatur: Raumtemperatur
MDL: MFCD01631407
CAS-Nummer: 58537-09-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR