2-Quinolin-4-yl-1-(4-trifluoromethylphenyl)ethanol 95+%

Lieferant: Apollo Scientific

PC0197-1G PC0197-25G PC0197-5G
APOSPC0197-1GEA 0 EUR
APOSPC0197-1G APOSPC0197-25G APOSPC0197-5G
2-Quinolin-4-yl-1-(4-trifluoromethylphenyl)ethanol 95+%
2-Quinolin-4-yl-1-(4-trifluoromethylphenyl)ethanol
Formel: C₁₈H₁₄F₃NO
Schmelzpunkt: 141…143 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD03424494
CAS-Nummer: 493024-38-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR