α-Brom-3-chlor-5-fluortoluol 98%

Lieferant: Apollo Scientific
Gefahr

Synonyme: 3-Chlor-5-fluorbenzylbromid

PC0198-5G PC0198-1G PC0198-25G
APOSPC0198-5GEA 37.7 EUR
APOSPC0198-5G APOSPC0198-1G APOSPC0198-25G
α-Brom-3-chlor-5-fluortoluol 98%
α-Brom-3-chlor-5-fluortoluol
Formel: C₇H₅BrClF
Molecular Weight: 223.47 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD03788472
CAS-Nummer: 493024-39-8
UN: 3265
ADR: 8 II

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR