5-Chloro-2-(trifluoromethyl)phenylacetonitrile 98%

Lieferant: Apollo Scientific

PC0204-10G PC0204-5G PC0204-1G
APOSPC0204-10GEA 0 EUR
APOSPC0204-10G APOSPC0204-5G APOSPC0204-1G
5-Chloro-2-(trifluoromethyl)phenylacetonitrile 98%
5-Chloro-2-(trifluoromethyl)phenylacetonitrile
Formel: C₉H₅ClF₃N
Schmelzpunkt: 61…63 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD01631357
CAS-Nummer: 261763-26-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR