2-Amino-6-fluorbenzamid 95+%
Lieferant: Apollo Scientific
|
Achtung
|
PC0205-5G
PC0205-25G
PC0205-100G
APOSPC0205-5GEA
43.5
EUR
APOSPC0205-5G
APOSPC0205-25G
APOSPC0205-100G
2-Amino-6-fluorbenzamid 95+%
2-Amino-6-fluorbenzamid
Formel:
C₇H₇FN₂O Molecular Weight: 154.1417 g/mol Schmelzpunkt: 115…118 °C Flammpunkt: >110 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00816881 CAS-Nummer: 115643-59-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...