(2,4-Dichlor-5-fluorphenyl)essigsäure 97%

Lieferant: Apollo Scientific

Synonyme: 2-(2,4-Dichloro-5-fluorophenyl)acetic acid

PC0215-1G
APOSPC0215-1GEA 132 EUR
APOSPC0215-1G
(2,4-Dichlor-5-fluorphenyl)essigsäure 97%
(2,4-Dichlor-5-fluorphenyl)essigsäure
Formel: (C₆H₂Cl₂F)CH₂COOH
Molecular Weight: 223.03 g/mol
Schmelzpunkt: 121…122 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD01631383
CAS-Nummer: 10129-77-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR