2,2-Difluor-1,3-benzodioxol-4-carbonylchlorid

Lieferant: Apollo Scientific
Gefahr

PC0219-1G
APOSPC0219-1GEA 212 EUR
APOSPC0219-1G
2,2-Difluor-1,3-benzodioxol-4-carbonylchlorid
2,2-Difluor-1,3-benzodioxol-4-carbonylchlorid
Formel: C₈H₃ClF₂O₃
Lagertemperatur: Raumtemperatur
MDL: MFCD01631474
CAS-Nummer: 143096-86-0
UN: 3265
ADR: 8,II

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR