3-(Trifluormethylthio)benzonitril 98%

Lieferant: Apollo Scientific
Achtung

PC0224-1G PC0224-250MG PC0224-5G
APOSPC0224-1GEA 325 EUR
APOSPC0224-1G APOSPC0224-250MG APOSPC0224-5G
3-(Trifluormethylthio)benzonitril 98%
3-(Trifluormethylthio)benzonitril
Formel: C₈H₄F₃NS
Lagertemperatur: Raumtemperatur
MDL: MFCD00236339
CAS-Nummer: 660-44-6
UN: 3276
ADR: 6.1 III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR