5-Phenyl-2-(trifluoromethyl)-3-furoic acid
Lieferant: Apollo Scientific
|
Achtung
|
PC0230-250MG
APOSPC0230-250MGEA
0
EUR
APOSPC0230-250MG
5-Phenyl-2-(trifluoromethyl)-3-furoic acid
5-Phenyl-2-(trifluoromethyl)-3-furoic acid
Formel:
C₁₂H₇F₃O₃ Schmelzpunkt: 164…166 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD01313502 CAS-Nummer: 23584-85-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...