N-Boc-2-fluoro-L-phenylalanine
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
Boc-Phe(2-F)-OH
PC0232-25G
APOSPC0232-25GEA
224
EUR
APOSPC0232-25G
N-Boc-2-fluoro-L-phenylalanine
N-Boc-2-fluoro-L-phenylalanine
Formel:
C₁₄H₁₈FNO₄ Molecular Weight: 283.299 g/mol Schmelzpunkt: 92…96 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00672520 CAS-Nummer: 114873-00-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...