3-(2H-Perfluoropropyl)adamantane-1-carboxylic acid 97%

Lieferant: Apollo Scientific
Achtung

Synonyme: 1-(1,1,2,3,3,3-Hexafluoroprop-1-yl)adamantane-3-carboxylic acid

PC0234-1G
APOSPC0234-1GEA 183 EUR
APOSPC0234-1G
3-(2H-Perfluoropropyl)adamantane-1-carboxylic acid 97%
3-(2H-Perfluoropropyl)adamantane-1-carboxylic acid
Formel: C₁₄H₁₆F₆O₂
Molecular Weight: 330,266 g/mol
Schmelzpunkt: 115…117 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00193108
CAS-Nummer: 86301-98-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR