8-Methoxy-3-[4-(trifluoromethyl)phenyl][1,2,4]triazolo[4,3-b]pyridazine 97%

Lieferant: Apollo Scientific
Achtung

Synonyme: 4-(8-Methoxy[1,2,4]triazolo[4,3-b]pyridazin-3-yl)benzotrifluoride

PC0235-100MG
APOSPC0235-100MGEA 49.3 EUR
APOSPC0235-100MG
8-Methoxy-3-[4-(trifluoromethyl)phenyl][1,2,4]triazolo[4,3-b]pyridazine 97%
8-Methoxy-3-[4-(trifluoromethyl)phenyl][1,2,4]triazolo[4,3-b]pyridazine
Formel: C₁₃H₉F₃N₄O
Molecular Weight: 294,232 g/mol
Schmelzpunkt: 142 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD01073588
CAS-Nummer: 230295-08-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR