3-(Trifluormethoxy)phenylboronsäure 98%
Lieferant: Apollo Scientific
|
Achtung
|
PC0240-25G
PC0240-5G
APOSPC0240-25GEA
0
EUR
APOSPC0240-25G
APOSPC0240-5G
3-(Trifluormethoxy)phenylboronsäure 98%
3-(Trifluormethoxy)phenylboronsäure
Formel:
C₇H₆BF₃O₃ Schmelzpunkt: 80…89 °C Lagertemperatur: Kühlschrank |
MDL:
MFCD01320697 CAS-Nummer: 179113-90-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...