2,3-Difluor-4-methylbenzamid 98%
Lieferant: Apollo Scientific
PC0241-1G
APOSPC0241-1GEA
27.7
EUR
APOSPC0241-1G
2,3-Difluor-4-methylbenzamid 98%
2,3-Difluor-4-methylbenzamid
Formel:
C₈H₇F₂NO Molecular Weight: 171,15 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD01631633 CAS-Nummer: 261763-35-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...