2,3-Difluor-p-kresol
Lieferant: Apollo Scientific
|
Gefahr
|
PC0259-1G
APOSPC0259-1GEA
21.9
EUR
APOSPC0259-1G
2,3-Difluor-p-kresol
2,3-Difluor-p-kresol
Formel:
C₇H₆F₂O Molecular Weight: 144,12 g/mol Schmelzpunkt: 35…38 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD01631651 CAS-Nummer: 261763-45-5 UN: 3261 ADR: 8,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...