2,6-Difluor-m-kresol
Lieferant: Apollo Scientific
|
Gefahr
|
PC0260-1G
APOSPC0260-1GEA
0
EUR
APOSPC0260-1G
2,6-Difluor-m-kresol
2,6-Difluor-m-kresol
Formel:
C₇H₆F₂O Siedepunkt: 182 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD01631340 CAS-Nummer: 261763-46-6 UN: 3261 ADR: 8,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...