(2,3-Difluor-4-methylphenyl)essigsäure
Lieferant: Apollo Scientific
|
Achtung
|
PC0261-250MG
APOSPC0261-250MGEA
111
EUR
APOSPC0261-250MG
(2,3-Difluor-4-methylphenyl)essigsäure
(2,3-Difluor-4-methylphenyl)essigsäure
Formel:
C₉H₈F₂O₂ Molecular Weight: 186,16 g/mol Schmelzpunkt: 124…126 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD01631649 CAS-Nummer: 261763-47-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...