2,3,6-Trifluorzimtsäure 98%

Lieferant: Apollo Scientific

Synonyme: 2,3,6-Trifluorcinnamsäure, 3-(2,3,6-Trifluorphenyl)propensäure

PC0275-5G PC0275-1G
APOSPC0275-5GEA 144 EUR
APOSPC0275-5G APOSPC0275-1G
2,3,6-Trifluorzimtsäure 98%
2,3,6-Trifluorzimtsäure
Formel: C₉H₅F₃O₂
Molecular Weight: 202.13 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00061213
CAS-Nummer: 230295-12-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR