2-(Difluoromethylthio)benzyl alcohol

Lieferant: Apollo Scientific

PC0279-1G
APOSPC0279-1GEA 364 EUR
APOSPC0279-1G
2-(Difluoromethylthio)benzyl alcohol
2-(Difluoromethylthio)benzyl alcohol
Formel: C₈H₈F₂OS
Lagertemperatur: Raumtemperatur
MDL: MFCD01631520
CAS-Nummer: 261944-18-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR