1,2-Difluorethylen (Gemisch der cis- und trans-Isomeren) 95%

Lieferant: Apollo Scientific
Achtung

Synonyme: FC-1132

PC0287-1G
APOSPC0287-1GEA 568 EUR
APOSPC0287-1G
1,2-Difluorethylen (Gemisch der cis- und trans-Isomeren) 95%
1,2-Difluorethylen (Gemisch der cis- und trans-Isomeren)

Lieferumfang: Delivery of this item to mainland UK is £35, additional charge will be added at order confirmation stage by our sales team.

Formel: C₂H₂F₂
Molecular Weight: 64,03 g/mol
Siedepunkt: –28 °C
CAS-Nummer: 1691-13-0
EINECS: 216-886-9
UN: 3161
ADR: 2.1,

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR