2-Brom-4,5-bis(trifluormethyl)anilin

Lieferant: Apollo Scientific
Achtung

Synonyme: 4,5-Bis(trifluoromethyl)-2-bromoaniline

PC0295-50MG
APOSPC0295-50MGEA 43.7 EUR
APOSPC0295-50MG
2-Brom-4,5-bis(trifluormethyl)anilin
2-Brom-4,5-bis(trifluormethyl)anilin
Formel: C₈H₄BrF₆N
Molecular Weight: 308,02 g/mol
Siedepunkt: 117…118 °C/12mmHg
Lagertemperatur: Raumtemperatur
CAS-Nummer: 230295-15-5
UN: 2811
ADR: 6.1,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR