5-(Piperidin-4-yl)-3-[3-(trifluoromethyl)phenyl]-1,2,4-oxadiazole 95%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
4-{3-[3-(Trifluoromethyl)phenyl]-1,2,4-oxadiazol-5-yl}piperidine
PC0299-250MG
APOSPC0299-250MGEA
180
EUR
APOSPC0299-250MG
5-(Piperidin-4-yl)-3-[3-(trifluoromethyl)phenyl]-1,2,4-oxadiazole 95%
5-(Piperidin-4-yl)-3-[3-(trifluoromethyl)phenyl]-1,2,4-oxadiazole
Formel:
C₁₄H₁₄F₃N₃O Molecular Weight: 297,28 g/mol Schmelzpunkt: 69…71 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD03424498 CAS-Nummer: 808764-45-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...