5,7-Difluoroxindol
Lieferant: Apollo Scientific
|
Achtung
|
PC0300-250MG
APOSPC0300-250MGEA
59.6
EUR
APOSPC0300-250MG
5,7-Difluoroxindol
5,7-Difluoroxindol
Formel:
C₈H₅F₂NO Schmelzpunkt: 220…222 °C Lagertemperatur: Kühlschrank |
MDL:
MFCD01075219 CAS-Nummer: 247564-59-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...