3-Fluor-4-(trifluormethyl)benzylalkohol 97%

Lieferant: Apollo Scientific
Achtung

Synonyme: (3-Fluoro-4-(trifluoromethyl)phenyl)methanol

PC0304-1G PC0304-5G PC0304-25G
APOSPC0304-1GEA 21.9 EUR
APOSPC0304-1G APOSPC0304-5G APOSPC0304-25G
3-Fluor-4-(trifluormethyl)benzylalkohol 97%
3-Fluor-4-(trifluormethyl)benzylalkohol
Formel: C₈H₆F₄O
Lagertemperatur: Raumtemperatur
MDL: MFCD00236285
CAS-Nummer: 230295-16-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR