3-(2-Fluorphenyl)propionsäure 98%
Lieferant: Apollo Scientific
|
Achtung
|
PC0312-1G
PC0312-25G
PC0312-5G
APOSPC0312-1GEA
58
EUR
APOSPC0312-1G
APOSPC0312-25G
APOSPC0312-5G
3-(2-Fluorphenyl)propionsäure 98%
3-(2-Fluorphenyl)propionsäure
Formel:
C₉H₉FO₂ Schmelzpunkt: 74…79 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD01310820 CAS-Nummer: 1643-26-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...