2-(4-Trifluoromethoxyphenyl)malondialdehyde 95%

Lieferant: Apollo Scientific
Achtung

PC0313-1G
APOSPC0313-1GEA 444 EUR
APOSPC0313-1G
2-(4-Trifluoromethoxyphenyl)malondialdehyde 95%
2-(4-Trifluoromethoxyphenyl)malondialdehyde
Formel: C₁₀H₇F₃O₃
Molecular Weight: 232,16 g/mol
Schmelzpunkt: 96…98 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD03424495
CAS-Nummer: 493036-48-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR