3-Chlor-4-(trifluormethyl)anilin 98+%

Lieferant: Apollo Scientific
Achtung

Synonyme: 4-Amino-2-chlor-α,α,α-trifluortoluol, 3-Chlor-α,α,α-trifluor-p-toluidin

PC0320-1G PC0320-25G PC0320-5G
APOSPC0320-1GEA 21.9 EUR
APOSPC0320-1G APOSPC0320-25G APOSPC0320-5G
3-Chlor-4-(trifluormethyl)anilin 98+%
3-Chlor-4-(trifluormethyl)anilin
Formel: C₇H₅ClF₃N
Molecular Weight: 195.57 g/mol
Siedepunkt: 200…202 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00278479
CAS-Nummer: 445-13-6
UN: 2810
ADR: 6.1 III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR