3-Chlor-4-(trifluormethyl)anilin 98+%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
4-Amino-2-chlor-α,α,α-trifluortoluol, 3-Chlor-α,α,α-trifluor-p-toluidin
PC0320-1G
PC0320-25G
PC0320-5G
APOSPC0320-1GEA
21.9
EUR
APOSPC0320-1G
APOSPC0320-25G
APOSPC0320-5G
3-Chlor-4-(trifluormethyl)anilin 98+%
3-Chlor-4-(trifluormethyl)anilin
Formel:
C₇H₅ClF₃N Molecular Weight: 195.57 g/mol Siedepunkt: 200…202 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00278479 CAS-Nummer: 445-13-6 UN: 2810 ADR: 6.1 III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...