4-Brom-3,5-difluorbenzotrifluorid 95%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
4-Brom-3,5-difluor-α,α,α-trifluortoluol, 1-Brom-2,6-difluor-4-(trifluormethyl)benzol
PC0321-1G
APOSPC0321-1GEA
250
EUR
APOSPC0321-1G
4-Brom-3,5-difluorbenzotrifluorid 95%
4-Brom-3,5-difluorbenzotrifluorid
Formel:
C₇H₂BrF₅ Molecular Weight: 260.99 g/mol Dichte: 1.74 g/cm³ (25 °C) Flammpunkt: 78 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD03093034 CAS-Nummer: 499238-36-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...