2-Chlor-4-nitrobenzotrifluorid
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
2-Chlor-4-nitrobenzotrifluorid, 1-Chlor-3-nitro-6-(trifluormethyl)benzol
PC0322-1G
PC0322-5G
APOSPC0322-1GEA
21.9
EUR
APOSPC0322-1G
APOSPC0322-5G
2-Chlor-4-nitrobenzotrifluorid
2-Chlor-4-nitrobenzotrifluorid
Formel:
C₇H₃ClF₃NO₂ Lagertemperatur: Raumtemperatur |
MDL:
MFCD00171532 CAS-Nummer: 151504-80-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...