7-Fluorisatin

Lieferant: Apollo Scientific
Achtung

Synonyme: 7-Fluorindolin-2,3-dion, 7-Fluor-2,3-indolindion, 7-Fluor-2,3-dioxoindolin, 7-Fluor-1H-indol-2,3-dion

PC0328-25G
APOSPC0328-25GEA 87 EUR
APOSPC0328-25G
7-Fluorisatin
7-Fluorisatin
Formel: C₈H₄FNO₂
Molecular Weight: 165,12 g/mol
Schmelzpunkt: 192…196 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD01569508
CAS-Nummer: 317-20-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR