Vinyl heptafluorobutanoate

Lieferant: Apollo Scientific
Gefahr

Synonyme: Vinyl perfluorobutanoate, Ethenyl heptafluorobutyrate

PC0337-10G
APOSPC0337-10GEA 223 EUR
APOSPC0337-10G
Vinyl heptafluorobutanoate
Vinyl heptafluorobutanoate
Formel: C₆H₃F₇O₂
Molecular Weight: 240,076 g/mol
Siedepunkt: 77…79 °C
Dichte: 1,41 g/cm³ (25 °C)
Lagertemperatur: Kühlschrank
MDL: MFCD09259000
CAS-Nummer: 356-28-5
UN: 1993
ADR: 3,II

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR