3'-Fluor-4'-methylacetophenon 98%
Lieferant: Apollo Scientific
|
Achtung
|
PC0338-25G
PC0338-5G
PC0338-1G
APOSPC0338-25GEA
393
EUR
APOSPC0338-25G
APOSPC0338-5G
APOSPC0338-1G
3'-Fluor-4'-methylacetophenon 98%
3'-Fluor-4'-methylacetophenon
Formel:
C₉H₉FO Schmelzpunkt: 31…34 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD01631411 CAS-Nummer: 42444-14-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...