5-Fluor-2-methylbenzylalkohol
Lieferant: Apollo Scientific
|
Achtung
|
PC0353-1G
PC0353-5G
PC0353-10G
APOSPC0353-1GEA
0
EUR
APOSPC0353-1G
APOSPC0353-5G
APOSPC0353-10G
5-Fluor-2-methylbenzylalkohol
5-Fluor-2-methylbenzylalkohol
Formel:
C₈H₉FO Schmelzpunkt: 36…40 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD01631527 CAS-Nummer: 22062-54-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...