3-Fluor-4-methylbenzylbromid

Lieferant: Apollo Scientific
Gefahr

Synonyme: 4-(Bromomethyl)-2-fluoro-1-methylbenzene

PC0360-10G PC0360-50G PC0360-5G
APOSPC0360-10GEA 116 EUR
APOSPC0360-10G APOSPC0360-50G APOSPC0360-5G
3-Fluor-4-methylbenzylbromid
3-Fluor-4-methylbenzylbromid
Formel: C₈H₈BrF
Molecular Weight: 203.05 g/mol
Siedepunkt: 77 °C/3mmHg
Lagertemperatur: Raumtemperatur
MDL: MFCD01631534
CAS-Nummer: 145075-44-1
UN: 3261
ADR: 8 III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR