2-Fluor-3-(trifluormethyl)anisol 98%

Lieferant: Apollo Scientific

PC0380-5G PC0380-1G
APOSPC0380-5GEA 0 EUR
APOSPC0380-5G APOSPC0380-1G
2-Fluor-3-(trifluormethyl)anisol 98%
2-Fluor-3-(trifluormethyl)anisol
Formel: C₈H₆F₄O
Lagertemperatur: Raumtemperatur
MDL: MFCD01631563
CAS-Nummer: 151868-17-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR