3-Fluor-5-(trifluormethyl)anisol 95%
Lieferant: Apollo Scientific
|
Achtung
|
PC0384-10G
PC0384-25G
PC0384-1G
APOSPC0384-10GEA
168
EUR
APOSPC0384-10G
APOSPC0384-25G
APOSPC0384-1G
3-Fluor-5-(trifluormethyl)anisol 95%
3-Fluor-5-(trifluormethyl)anisol
Formel:
C₈H₆F₄O Flammpunkt: 52 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD01631491 CAS-Nummer: 261951-79-5 UN: 1993 ADR: 3,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...