3-Fluor-2-(trifluormethyl)benzonitril 98%

Lieferant: Apollo Scientific
Achtung

PC0390-1G PC0390-5G PC0390-10G
APOSPC0390-1GEA 21.9 EUR
APOSPC0390-1G APOSPC0390-5G APOSPC0390-10G
3-Fluor-2-(trifluormethyl)benzonitril 98%
3-Fluor-2-(trifluormethyl)benzonitril
Formel: C₈H₃F₄N
Molecular Weight: 189.11 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD01631566
CAS-Nummer: 261951-81-9

Bestellen

TECHNISCHE DATEN

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR