3-Fluoro-2-(trifluoromethyl)benzoyl chloride 97%

Lieferant: Apollo Scientific
Gefahr

PC0391-1G PC0391-5G PC0391-10G
APOSPC0391-1GEA 0 EUR
APOSPC0391-1G APOSPC0391-5G APOSPC0391-10G
3-Fluoro-2-(trifluoromethyl)benzoyl chloride 97%
3-Fluoro-2-(trifluoromethyl)benzoyl chloride
Formel: C₈H₃ClF₄O
Lagertemperatur: Raumtemperatur
MDL: MFCD01631458
CAS-Nummer: 261951-82-0
UN: 1760
ADR: 8,III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR