4-Methoxy-3-(trifluormethyl)benzonitril

Lieferant: Apollo Scientific
Achtung

PC0400-25G PC0400-5G PC0400-100G
APOSPC0400-25GEA 145 EUR
APOSPC0400-25G APOSPC0400-5G APOSPC0400-100G
4-Methoxy-3-(trifluormethyl)benzonitril
4-Methoxy-3-(trifluormethyl)benzonitril
Formel: C₉H₆F₃NO
Molecular Weight: 201.15 g/mol
Schmelzpunkt: 98…100 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD01631575
CAS-Nummer: 261951-87-5
UN: 3439
ADR: 6.1 III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR