(Bis(trifluoracetoxy)iod)benzol 97%

Lieferant: Apollo Scientific
Achtung

PC0403-100G PC0403-25G
APOSPC0403-100GEA 87 EUR
APOSPC0403-100G APOSPC0403-25G
(Bis(trifluoracetoxy)iod)benzol 97%
(Bis(trifluoracetoxy)iod)benzol

Used to estimate asparagine and glutamine in peptides and proteins.

Formel: C₁₀H₅F₆IO₄
Schmelzpunkt: 122…125 °C
Lagertemperatur: Kühlschrank
MDL: MFCD00009672
CAS-Nummer: 2712-78-9
EINECS: 220-308-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR