N-(tert-Butyldimethylsilyl)-N-methyltrifluoracetamid (MTBSTFA) 97%

Lieferant: Apollo Scientific
Achtung

PC0406-100G PC0406-25G PC0406-5G
APOSPC0406-100GEA 241 EUR
APOSPC0406-100G APOSPC0406-25G APOSPC0406-5G
N-(tert-Butyldimethylsilyl)-N-methyltrifluoracetamid (MTBSTFA) 97%
N-(tert-Butyldimethylsilyl)-N-methyltrifluoracetamid (MTBSTFA)

GC derivatising agent for silylation of hydroxy, carboxy, thio & primary and secondary amines.

Formel: C₉H₁₈F₃NOSi
Siedepunkt: 168…170 °C
Dichte: 1,023 g/cm³ (25 °C)
Flammpunkt: 52 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00009671
CAS-Nummer: 77377-52-7
UN: 1993
ADR: 3,III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR