4-Methoxy-3-(trifluoromethyl)benzyl alcohol 97%

Lieferant: Apollo Scientific
Achtung

PC0410-5G
APOSPC0410-5GEA 0 EUR
APOSPC0410-5G
4-Methoxy-3-(trifluoromethyl)benzyl alcohol 97%
4-Methoxy-3-(trifluoromethyl)benzyl alcohol
Formel: C₉H₉F₃O₂
Lagertemperatur: Raumtemperatur
MDL: MFCD01631577
CAS-Nummer: 261951-88-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR